Assay Details
| Gene Symbol: sri-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276854.1 | NP_001263783.1 | 5 - 6 | 679 | 135 | |
| NM_069642.1 | NP_502043.1 | 5 - 6 | 832 | 135 | ||
| NR_131607.1 | 6 - 7 | 865 | 135 | |||
More Information
Set Membership: |
|

sri-19
Genomic Map