Assay Details
| Gene Symbol: aqp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306967.3 | NP_001293896.1 | 3 - 4 | 918 | 143 | |
| NM_001322776.4 | NP_001309613.1 | 5 - 6 | 1118 | 143 | ||
| NM_069643.2 | NP_502044.1 | 5 - 6 | 1215 | 143 | ||
More Information
Set Membership: |
|

aqp-3
Genomic Map