Assay Details
| Gene Symbol: alh-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069653.9 | NP_502054.2 | 4 - 5 | 1157 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

alh-3| Gene Symbol: alh-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069653.9 | NP_502054.2 | 4 - 5 | 1157 | 73 | |
Set Membership: |
|