Assay Details
| Gene Symbol: klc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268605.3 | NP_001255534.1 | 2 - 3 | 182 | 117 | |
| NM_001268606.4 | NP_001255535.1 | 1 - 2 | 145 | 117 | ||
More Information
Set Membership: |
|

klc-1
Genomic Map