Assay Details
| Gene Symbol: spo-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307081.3 | NP_001294010.1 | 1 - 2 | 76 | 125 | |
| NM_069680.5 | NP_502081.1 | 2 - 3 | 344 | 125 | ||
More Information
Set Membership: |
|

spo-11
Genomic Map