Assay Details
| Gene Symbol: spo-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307081.3 | NP_001294010.1 | 4 - 5 | 633 | 140 | |
| NM_069680.5 | NP_502081.1 | 5 - 6 | 901 | 140 | ||
More Information
Set Membership: |
|

spo-11
Genomic Map