Assay Details
| Gene Symbol: swsn-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069681.7 | NP_502082.1 | 7 - 8 | 4397 | 91 | |
| GenBank mRNA | AF230280.1 | - | 7 - 8 | 4397 | 91 | |
More Information
Set Membership: |
|

swsn-4
Genomic Map