Assay Details
| Gene Symbol: cpt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047512.4 | NP_001040977.2 | 6 - 7 | 996 | 99 | |
| NM_001047513.5 | NP_001040978.2 | 4 - 5 | 486 | 99 | ||
More Information
Set Membership: |
|

cpt-2
Genomic Map