Assay Details
| Gene Symbol: cpt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047512.4 | NP_001040977.2 | 9 - 10 | 1855 | 94 | |
| NM_001047513.5 | NP_001040978.2 | 7 - 8 | 1345 | 94 | ||
| NM_001307075.3 | NP_001294004.1 | 1 - 2 | 244 | 94 | ||
More Information
Set Membership: |
|

cpt-2
Genomic Map