Assay Details
| Gene Symbol: gtl-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069717.7 | NP_502118.4 | 17 - 18 | 3501 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gtl-2| Gene Symbol: gtl-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069717.7 | NP_502118.4 | 17 - 18 | 3501 | 69 | |
Set Membership: |
|