Assay Details
| Gene Symbol: znf-207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069723.5 | NP_502124.1 | 4 - 5 | 736 | 70 | |
| NM_069724.4 | NP_502125.1 | 4 - 5 | 736 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

znf-207| Gene Symbol: znf-207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069723.5 | NP_502124.1 | 4 - 5 | 736 | 70 | |
| NM_069724.4 | NP_502125.1 | 4 - 5 | 736 | 70 | ||
Set Membership: |
|