Assay Details
| Gene Symbol: leo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069734.4 | NP_502135.1 | 2 - 3 | 459 | 116 | |
| Gene Symbol: sart-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069735.7 | NP_502136.1 | 12 - 13 | 3389 | 116 | |
More Information
Set Membership: |
|

leo-1
Genomic Map