Assay Details
| Gene Symbol: leo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069734.4 | NP_502135.1 | 3 - 4 | 627 | 73 | |
| Gene Symbol: sart-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069735.7 | NP_502136.1 | 13 - 14 | 3557 | 73 | |
More Information
Set Membership: |
|

leo-1
Genomic Map