Assay Details
| Gene Symbol: leo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069734.4 | NP_502135.1 | 5 - 6 | 822 | 95 | |
| Gene Symbol: sart-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069735.7 | NP_502136.1 | 15 - 16 | 3752 | 95 | |
More Information
Set Membership: |
|

leo-1
Genomic Map