Assay Details
| Gene Symbol: leo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069734.4 | NP_502135.1 | 6 - 7 | 1153 | 77 | |
| Gene Symbol: sart-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069735.7 | NP_502136.1 | 16 - 17 | 4083 | 77 | |
More Information
Set Membership: |
|

leo-1
Genomic Map