Assay Details
Gene Symbol: K11H12.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067562.5 | NP_499963.1 | 3 - 4 | 808 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: K11H12.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067562.5 | NP_499963.1 | 3 - 4 | 808 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() |