Assay Details
| Gene Symbol: rpl-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067563.7 | NP_499964.1 | 2 - 3 | 316 | 72 | |
| Gene Symbol: K11H12.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067562.5 | NP_499963.1 | 2 - 3 | 301 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_067562.5 transcripts.
Set Membership: |
|

rpl-15
Genomic Map