Assay Details
| Gene Symbol: mak-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380025.1 | NP_001367654.1 | 2 - 3 | 252 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380025.1 transcripts.
Set Membership: |
|

mak-2
Genomic Map