Assay Details
| Gene Symbol: faah-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067616.3 | NP_500017.2 | 4 - 5 | 673 | 149 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

faah-3| Gene Symbol: faah-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067616.3 | NP_500017.2 | 4 - 5 | 673 | 149 | |
Set Membership: |
|