Assay Details
| Gene Symbol: plx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307625.3 | NP_001294554.1 | 5 - 6 | 933 | 70 | |
| NM_001307626.3 | NP_001294555.1 | 2 - 3 | 366 | 70 | ||
| NM_001307627.3 | NP_001294556.1 | 2 - 3 | 366 | 70 | ||
| NM_067617.6 | NP_500018.3 | 5 - 6 | 933 | 70 | ||
More Information
Set Membership: |
|

plx-1
Genomic Map