Assay Details
| Gene Symbol: cct-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307548.3 | NP_001294477.1 | 3 - 4 | 1092 | 101 | |
| NM_067634.5 | NP_500035.3 | 6 - 7 | 1462 | 101 | ||
More Information
Set Membership: |
|

cct-8
Genomic Map