Assay Details
| Gene Symbol: ost-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380031.1 | NP_001368314.1 | 1 - 2 | 212 | 60 | |
| NM_067638.8 | NP_500039.1 | 4 - 5 | 526 | 60 | ||
More Information
Set Membership: |
|

ost-1
Genomic Map