Assay Details
| Gene Symbol: ost-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380031.1 | NP_001368314.1 | 2 - 3 | 364 | 92 | |
| NM_067638.8 | NP_500039.1 | 5 - 6 | 678 | 92 | ||
More Information
Set Membership: |
|

ost-1
Genomic Map