Assay Details
| Gene Symbol: clp-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380044.1 | NP_001368280.1 | 1 - 2 | 146 | 110 | |
| NM_067681.6 | NP_500082.2 | 5 - 6 | 1607 | 110 | ||
More Information
Set Membership: |
|

clp-7
Genomic Map