Assay Details
| Gene Symbol: clp-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380044.1 | NP_001368280.1 | 2 - 3 | 395 | 70 | |
| NM_067681.6 | NP_500082.2 | 6 - 7 | 1856 | 70 | ||
More Information
Set Membership: |
|

clp-7
Genomic Map