Assay Details
| Gene Symbol: Y41D4A.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067730.7 | NP_500131.3 | 2 - 3 | 1084 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Y41D4A.3| Gene Symbol: Y41D4A.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067730.7 | NP_500131.3 | 2 - 3 | 1084 | 65 | |
Set Membership: |
|