Assay Details
| Gene Symbol: cpsf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067756.6 | NP_500157.2 | 4 - 5 | 652 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpsf-1| Gene Symbol: cpsf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067756.6 | NP_500157.2 | 4 - 5 | 652 | 137 | |
Set Membership: |
|