Assay Details
| Gene Symbol: cpsf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380077.1 | NP_001368311.1 | 4 - 5 | 1141 | 89 | |
| NM_067756.6 | NP_500157.2 | 7 - 8 | 1718 | 89 | ||
More Information
Set Membership: |
|

cpsf-1
Genomic Map