Assay Details
| Gene Symbol: cpsf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380077.1 | NP_001368311.1 | 5 - 6 | 1620 | 118 | |
| NM_067756.6 | NP_500157.2 | 8 - 9 | 2197 | 118 | ||
More Information
Set Membership: |
|

cpsf-1
Genomic Map