Assay Details
Gene Symbol: prmt-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067767.5 | NP_500168.2 | 2 - 3 | 617 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: prmt-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067767.5 | NP_500168.2 | 2 - 3 | 617 | 87 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |