Assay Details
| Gene Symbol: gcy-37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067770.5 | NP_500171.2 | 3 - 4 | 232 | 111 | |
| GenBank mRNA | AY652946.1 | - | 3 - 4 | 225 | 111 | |
More Information
Set Membership: |
|

gcy-37
Genomic Map