Assay Details
| Gene Symbol: cdh-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067803.6 | NP_500204.3 | 16 - 17 | 4885 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdh-10| Gene Symbol: cdh-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067803.6 | NP_500204.3 | 16 - 17 | 4885 | 92 | |
Set Membership: |
|