Assay Details
| Gene Symbol: smf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067834.6 | NP_500235.4 | 11 - 12 | 1684 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

smf-3| Gene Symbol: smf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067834.6 | NP_500235.4 | 11 - 12 | 1684 | 77 | |
Set Membership: |
|