Assay Details
| Gene Symbol: dib-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276811.3 | NP_001263740.1 | 3 - 4 | 515 | 70 | |
| Gene Symbol: dml-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028334.7 | NP_001023505.2 | 5 - 6 | 1103 | 70 | |
More Information
Set Membership: |
|

dib-1
Genomic Map