Assay Details
| Gene Symbol: lad-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028325.3 | NP_001023496.1 | 11 - 12 | 1798 | 75 | |
| GenBank mRNA | AF456361.1 | - | 11 - 12 | 1802 | 75 | |
| EF584655.1 | - | 11 - 12 | 1712 | 75 | ||
More Information
Set Membership: |
|

lad-2
Genomic Map