Assay Details
| Gene Symbol: lad-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028325.3 | NP_001023496.1 | 3 - 4 | 380 | 151 | |
| NM_001028326.4 | NP_001023497.1 | 3 - 4 | 294 | 151 | ||
| GenBank mRNA | AF456361.1 | - | 3 - 4 | 384 | 151 | |
| EF584655.1 | - | 3 - 4 | 294 | 151 | ||
More Information
Set Membership: |
|

lad-2
Genomic Map