Assay Details
| Gene Symbol: dmd-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067904.4 | NP_500305.1 | 3 - 4 | 673 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dmd-9| Gene Symbol: dmd-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067904.4 | NP_500305.1 | 3 - 4 | 673 | 72 | |
Set Membership: |
|