Assay Details
| Gene Symbol: T26C12.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067905.5 | NP_500306.1 | 3 - 4 | 1638 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

T26C12.1| Gene Symbol: T26C12.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067905.5 | NP_500306.1 | 3 - 4 | 1638 | 79 | |
Set Membership: |
|