Assay Details
| Gene Symbol: gcy-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067908.5 | NP_500309.3 | 6 - 7 | 2432 | 131 | |
| GenBank mRNA | AB201390.1 | - | 6 - 7 | 2432 | 131 | |
More Information
Set Membership: |
|

gcy-23
Genomic Map