Assay Details
| Gene Symbol: zip-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067917.6 | NP_500318.1 | 1 - 2 | 334 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

zip-11| Gene Symbol: zip-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067917.6 | NP_500318.1 | 1 - 2 | 334 | 126 | |
Set Membership: |
|