Assay Details
| Gene Symbol: elks-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067928.4 | NP_500329.1 | 4 - 5 | 672 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

elks-1| Gene Symbol: elks-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067928.4 | NP_500329.1 | 4 - 5 | 672 | 103 | |
Set Membership: |
|