Assay Details
| Gene Symbol: srz-79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067958.4 | NP_500359.1 | 3 - 4 | 560 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srz-79| Gene Symbol: srz-79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067958.4 | NP_500359.1 | 3 - 4 | 560 | 153 | |
Set Membership: |
|