Assay Details
| Gene Symbol: tsg-101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067963.5 | NP_500364.2 | 3 - 4 | 639 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tsg-101| Gene Symbol: tsg-101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067963.5 | NP_500364.2 | 3 - 4 | 639 | 94 | |
Set Membership: |
|