Assay Details
| Gene Symbol: osm-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067971.4 | NP_500372.1 | 11 - 12 | 2064 | 67 | |
| GenBank mRNA | AF031408.1 | - | 11 - 12 | 2064 | 67 | |
More Information
Set Membership: |
|

osm-9
Genomic Map