Assay Details
| Gene Symbol: osm-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067971.4 | NP_500372.1 | 4 - 5 | 516 | 82 | |
| GenBank mRNA | AF031408.1 | - | 4 - 5 | 516 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

osm-9| Gene Symbol: osm-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067971.4 | NP_500372.1 | 4 - 5 | 516 | 82 | |
| GenBank mRNA | AF031408.1 | - | 4 - 5 | 516 | 82 | |
Set Membership: |
|