Assay Details
| Gene Symbol: cdc-73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068064.7 | NP_500465.4 | 6 - 7 | 934 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdc-73| Gene Symbol: cdc-73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068064.7 | NP_500465.4 | 6 - 7 | 934 | 100 | |
Set Membership: |
|