Assay Details
| Gene Symbol: pax-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307721.3 | NP_001294650.1 | 6 - 7 | 683 | 74 | |
| NM_068112.4 | NP_500513.1 | 7 - 8 | 869 | 74 | ||
More Information
Set Membership: |
|

pax-2
Genomic Map