Assay Details
| Gene Symbol: sars-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068148.8 | NP_500549.1 | 1 - 2 | 188 | 103 | |
| GenBank mRNA | AB016796.1 | - | 1 - 2 | 191 | 103 | |
More Information
Set Membership: |
|

sars-2
Genomic Map