Assay Details
| Gene Symbol: sars-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380185.1 | NP_001368218.1 | 1 - 2 | 326 | 71 | |
| NM_068148.8 | NP_500549.1 | 3 - 4 | 979 | 71 | ||
| GenBank mRNA | AB016796.1 | - | 3 - 4 | 982 | 71 | |
More Information
Set Membership: |
|

sars-2
Genomic Map