Assay Details
| Gene Symbol: chat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028161.4 | NP_001023332.1 | 2 - 3 | 405 | 62 | |
| NR_138505.1 | 2 - 3 | 404 | 62 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

chat-1| Gene Symbol: chat-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028161.4 | NP_001023332.1 | 2 - 3 | 405 | 62 | |
| NR_138505.1 | 2 - 3 | 404 | 62 | |||
Set Membership: |
|